siRNA Details
Gene Symbol: Sv2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057207.3 | NP_476555.1 | 2 | 525 | ||
XM_008759492.1 | XP_008757714.1 | 2 | 1244 | |||
XM_008759493.1 | XP_008757715.1 | 2 | 1622 | |||
XM_008759494.1 | XP_008757716.1 | 2 | 2144 | |||
XM_008759495.1 | XP_008757717.1 | 2 | 1240 | |||
GenBank mRNA | L10362.1 | - | 2 | 525 |
