siRNA Details
Gene Symbol: Sv2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057207.3 | NP_476555.1 | 13 | 2786 | ||
XM_008759492.1 | XP_008757714.1 | 13 | 3505 | |||
XM_008759493.1 | XP_008757715.1 | 13 | 3883 | |||
XM_008759494.1 | XP_008757716.1 | 13 | 4405 | |||
XM_008759495.1 | XP_008757717.1 | 13 | 3501 | |||
GenBank mRNA | CB609053.1 | - | 1 | 223 | ||
L10362.1 | - | 13 | 2786 |
