siRNA Details
Gene Symbol: Tpm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301285.1 | NP_001288214.1 | 6 | 696 | ||
NM_001301286.1 | NP_001288215.1 | 5 | 520 | |||
NM_057208.2 | NP_476556.2 | 5 | 520 | |||
XM_006232572.2 | XP_006232634.1 | 6 | 716 | |||
XM_006232573.2 | XP_006232635.1 | 6 | 716 | |||
XM_006232575.2 | XP_006232637.1 | 6 | 716 | |||
XM_006232577.2 | XP_006232639.1 | 6 | 716 | |||
XM_006232581.2 | XP_006232643.1 | 5 | 621 | |||
XM_006232583.2 | XP_006232645.1 | 5 | 624 | |||
GenBank mRNA | AF053361.1 | - | 2 | 125 | ||
FQ214881.1 | - | 6 | 694 | |||
FQ215580.1 | - | 6 | 696 | |||
FQ215752.1 | - | 6 | 696 | |||
FQ216429.1 | - | 6 | 695 | |||
L24776.1 | - | 520 | ||||
L24777.1 | - | 5 | 520 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
