siRNA Details
Gene Symbol: UGT1A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007120.2 | NP_009051.1 | 1 | 298 | ||
GenBank mRNA | AK313623.1 | - | 1 | 312 | ||
AY435139.1 | - | 1 | 269 | |||
BC121036.1 | - | 1 | 269 | 40117585 | ||
BC131623.1 | - | 1 | 262 | |||
BC139784.1 | - | 1 | 289 | 40117089 | ||
DQ364249.1 | - | 1 | 278 | |||
M57951.1 | - | 1 | 298 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
