siRNA Details
Gene Symbol: Gata3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133293.1 | NP_579827.1 | 1 | 152 | ||
XM_006254253.2 | XP_006254315.1 | 2 | 918 | |||
XM_008771893.1 | XP_008770115.1 | 2 | 862 | |||
XM_008771894.1 | XP_008770116.1 | 2 | 2452 | |||
GenBank mRNA | AB000217.1 | - | 1 | 363 | ||
AY024364.1 | - | 1 | 152 |

More Information
Other Species Targeted

Gene Ontology Categories:
