siRNA Details
Gene Symbol: Zfp111 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133323.1 | NP_579857.1 | 3 | 421 | ||
XM_006228375.2 | XP_006228437.1 | 1 | 361 | |||
XM_006228376.2 | XP_006228438.1 | 2 | 517 | |||
GenBank mRNA | FQ211918.1 | - | 3 | 487 | ||
U27186.1 | - | 3 | 421 |
Gene Symbol: LOC102548230 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006228505.2 | XP_006228567.1 | 2 | 1499 | ||
XM_006228506.2 | XP_006228568.1 | 3 | 755 | |||
XM_006228507.2 | XP_006228569.1 | 3 | 634 |
Gene Symbol: LOC102548503 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759056.1 | XP_008757278.1 | 3 | 434 |
Gene Symbol: LOC102548600 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759057.1 | XP_008757279.1 | 3 | 222 |
Gene Symbol: Zfp108 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_574384.6 | XP_574384.4 | 1 | 203 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
