siRNA Details
Gene Symbol: Sphk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB049571.1 | - | 6 | 1767 | ||
AB049572.1 | - | 6 | 1719 | |||
AB049573.1 | - | 6 | 1666 | |||
AB049574.1 | - | 6 | 2325 | |||
AB049575.1 | - | 5 | 2472 | |||
DQ486894.1 | - | 6 | 1507 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
