siRNA Details
Gene Symbol: ZNF184 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK312804.1 | - | 3, 4 | 609 | ||
BC022992.1 | - | 3, 4 | 350 | 4997766 | ||
DB090589.1 | - | 3, 4 | 368 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
