siRNA Details
Gene Symbol: ADORA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000677.3 | NP_000668.1 | 2 | 1815 | ||
NM_001302678.1 | NP_001289607.1 | 2 | 1879 | |||
NM_001302679.1 | NP_001289608.1 | 2 | 1321 | |||
GenBank mRNA | CD013875.1 | - | 2 | 1170 | ||
JN855714.1 | - | 2 | 1047 | |||
L20463.1 | - | 2 | 1339 | |||
L22607.1 | - | 2 | 1313 | |||
X76981.1 | - | 2 | 1381 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
