siRNA Details
Gene Symbol: CSDE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007553.2 | NP_001007554.1 | 5 | 897 | ||
NM_001242891.1 | NP_001229820.1 | 6 | 1044 | |||
NM_001242892.1 | NP_001229821.1 | 4 | 510 | |||
GenBank mRNA | AB020692.1 | - | 5 | 811 | ||
AK290146.1 | - | 4 | 442 | |||
CR749378.1 | - | 5 | 901 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
