siRNA Details
Gene Symbol: Pmfbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134393.2 | NP_599220.1 | 3 | 273 | ||
XM_006255570.2 | XP_006255632.2 | 4 | 1017 | |||
XM_006255571.2 | XP_006255633.2 | 4 | 1017 | |||
XM_006255572.2 | XP_006255634.1 | 4 | 1331 | |||
XM_006255573.2 | XP_006255635.1 | 4 | 572 | |||
XM_006255574.2 | XP_006255636.2 | 4 | 1015 | |||
XM_006255576.2 | XP_006255638.2 | 4 | 1016 | |||
XM_008772468.1 | XP_008770690.1 | 3 | 861 | |||
XM_008772469.1 | XP_008770691.1 | 3 | 415 | |||
XM_008772470.1 | XP_008770692.1 | 4 | 1017 | |||
GenBank mRNA | AF092090.2 | - | 3 | 278 |
