siRNA Details
Gene Symbol: Akr1d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138884.1 | NP_620239.1 | 1 | 122 | ||
GenBank mRNA | D17309.1 | - | 1 | 122 | ||
FQ211413.1 | - | 1 | 140 | |||
FQ219185.1 | - | 1 | 141 | |||
FQ219421.1 | - | 1 | 85 | |||
FQ219730.1 | - | 1 | 85 | |||
S80431.1 | - | 1 | 94 |
