siRNA Details
Gene Symbol: Fbxo6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138917.3 | NP_620272.3 | 3 | 450 | ||
XM_006239372.2 | XP_006239434.1 | 4 | 754 | |||
XM_006239373.2 | XP_006239435.1 | 4 | 687 | |||
XM_006239374.2 | XP_006239436.1 | 4 | 614 | |||
XM_006239375.2 | XP_006239437.1 | 4 | 618 | |||
GenBank mRNA | AF393484.1 | - | 3 | 450 | ||
FQ232698.1 | - | 4 | 520 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
