siRNA Details
Gene Symbol: Ppp1r14b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172045.1 | NP_742042.1 | 2, 3 | 361 | ||
GenBank mRNA | AY122323.1 | - | 2, 3 | 361 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
