siRNA Details
Gene Symbol: HHLA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282556.1 | NP_001269485.1 | 4 | 441 | ||
NM_001282557.1 | NP_001269486.1 | 4 | 372 | |||
NM_001282558.1 | NP_001269487.1 | 5 | 519 | |||
NM_007072.3 | NP_009003.1 | 4 | 632 | |||
XM_005247080.3 | XP_005247137.1 | 5 | 502 | |||
XM_011512362.1 | XP_011510664.1 | 5 | 502 | |||
XM_011512363.1 | XP_011510665.1 | 4 | 432 | |||
XM_011512364.1 | XP_011510666.1 | 4 | 383 | |||
XM_011512367.2 | XP_011510669.1 | 5 | 502 | |||
XM_017005642.1 | XP_016861131.1 | 4 | 432 | |||
GenBank mRNA | AF126162.1 | - | 4 | 372 | ||
AK000692.1 | - | 3 | 367 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
