siRNA Details
Gene Symbol: Spg7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181388.2 | NP_852053.1 | 2 | 279 | ||
XM_008772619.1 | XP_008770841.1 | 2 | 286 | |||
XM_008772620.1 | XP_008770842.1 | 2 | 263 | |||
GenBank mRNA | AY278739.1 | - | 2 | 279 | ||
FQ224821.1 | - | 2 | 291 |
