siRNA Details
Gene Symbol: Nsmaf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181389.2 | NP_852054.1 | 2 | 121 | ||
XM_008763544.1 | XP_008761766.1 | 2 | 1180 | |||
XM_008763545.1 | XP_008761767.1 | 2 | 1180 | |||
GenBank mRNA | AY163240.1 | - | 2 | 121 |
