siRNA Details
Gene Symbol: Col23a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181636.2 | NP_853667.1 | 2642 | |||
GenBank mRNA | AY158896.1 | - | 2642 |
Gene Symbol: LOC103693323 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008767739.1 | XP_008765961.1 | 2026 |
