siRNA Details
Gene Symbol: Tbx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181638.1 | NP_853669.1 | 2 | 637 | ||
XM_006249415.2 | XP_006249477.1 | 3 | 1785 | |||
XM_006249416.2 | XP_006249478.1 | 3 | 2648 | |||
XM_006249417.2 | XP_006249479.1 | 3 | 1795 | |||
XM_006249418.1 | XP_006249480.1 | 2 | 1909 | |||
GenBank mRNA | AY289619.1 | - | 2 | 637 | ||
FQ221880.1 | - | 2 | 1607 |
