siRNA Details
Gene Symbol: Nit1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082580.1 | NP_001076049.1 | 6 | 800 | ||
NM_182668.2 | NP_872609.2 | 6 | 768 | |||
GenBank mRNA | AY300752.1 | - | 5 | 631 | ||
BC087146.1 | - | 6 | 749 | 7129801 | ||
FQ213884.1 | - | 551 | ||||
FQ214471.1 | - | 798 | ||||
FQ227281.1 | - | 797 |
