siRNA Details
Gene Symbol: Nit1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082580.1 | NP_001076049.1 | 3 | 442 | ||
NM_182668.2 | NP_872609.2 | 3 | 410 | |||
GenBank mRNA | AY300752.1 | - | 2 | 273 | ||
BC087146.1 | - | 3 | 391 | 7129801 | ||
CB576182.1 | - | 3 | 505 | |||
CO558072.1 | - | 3 | 448 | |||
FQ213884.1 | - | 297 | ||||
FQ214471.1 | - | 440 | ||||
FQ227281.1 | - | 439 |
