siRNA Details
Gene Symbol: Arl6ip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198737.2 | NP_942032.1 | 3 | 328 | ||
GenBank mRNA | AY316590.1 | - | 3 | 329 | ||
BC099071.1 | - | 3 | 328 | 7381266 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
