siRNA Details
Gene Symbol: Atp1a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012505.2 | NP_036637.1 | 3 | 244 | ||
GenBank mRNA | BC085764.1 | - | 3 | 244 | 7190910 | |
M14512.1 | - | 3 | 229 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_178405, AK137602, BC036127, AK168703, AK147331, and others.
Gene Ontology Categories:
