siRNA Details
Gene Symbol: Atp1a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012505.2 | NP_036637.1 | 23 | 3492 | ||
GenBank mRNA | M14512.1 | - | 23 | 3477 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Atp1a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012505.2 | NP_036637.1 | 23 | 3492 | ||
GenBank mRNA | M14512.1 | - | 23 | 3477 |