siRNA Details
Gene Symbol: Hbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013221.2 | NP_037353.2 | 2, 3 | 219 | ||
XM_006239975.2 | XP_006240037.1 | 2, 3 | 282 | |||
GenBank mRNA | BC091165.1 | - | 1, 2 | 193 | 7097013 | |
FQ223257.1 | - | 2, 3 | 326 | |||
FQ224288.1 | - | 2, 3 | 210 | |||
U09551.1 | - | 1, 2 | 214 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_177993, NM_153198, AK053005, AK142187, AK133127, and others.
Gene Ontology Categories:
