siRNA Details
Gene Symbol: Syne1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006227853.2 | XP_006227915.1 | 17 | 2282 | ||
XM_008758714.1 | XP_008756936.1 | 16 | 2268 | |||
XM_008758715.1 | XP_008756937.1 | 18 | 2335 | |||
XM_008758716.1 | XP_008756938.1 | 18 | 2333 | |||
GenBank mRNA | AY597251.1 | - | 4 | 678 | ||
X95466.1 | - | 3 | 590 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
