siRNA Details
Gene Symbol: ZXDB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007157.3 | NP_009088.1 | 1 | 2689 | ||
GenBank mRNA | AK124513.1 | - | 2 | 688 | ||
AK289766.1 | - | 1 | 2499 |
Gene Symbol: ZXDA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007156.4 | NP_009087.1 | 1 | 2677 | ||
GenBank mRNA | AK122815.1 | - | 1 | 2676 | ||
AK292686.1 | - | 1 | 2667 | |||
BC059356.1 | - | 1 | 2475 | 30347480 | ||
L14787.1 | - | 1 | 1559 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
