siRNA Details
Gene Symbol: Tmem33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021671.3 | NP_067703.1 | 8 | 1392 | ||
GenBank mRNA | AB006135.1 | - | 8 | 1350 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
