siRNA Details
Gene Symbol: Rapgef3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021690.1 | NP_067722.1 | 3 | 384 | ||
XM_006242352.2 | XP_006242414.1 | 4 | 644 | |||
XM_008765778.1 | XP_008764000.1 | 4 | 933 | |||
XM_008765779.1 | XP_008764001.1 | 4 | 551 | |||
XM_008765780.1 | XP_008764002.1 | 3 | 891 | |||
GenBank mRNA | BC107667.1 | - | 3 | 337 | 7936411 | |
U78167.1 | - | 3 | 384 |
