siRNA Details
Gene Symbol: Ppp2r2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC078834.1 | - | 4, 5 | 487 | 7114294 | |
D14421.1 | - | 1, 2 | 53 | |||
D38260.1 | - | 1, 2 | 73 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
