siRNA Details
Gene Symbol: TOPBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007027.3 | NP_008958.2 | 10 | 1440 | ||
XM_005247076.3 | XP_005247133.1 | 10 | 1912 | |||
XM_011512357.2 | XP_011510659.1 | 10 | 1511 | |||
XM_017005636.1 | XP_016861125.1 | 10 | 1510 | |||
XM_017005637.1 | XP_016861126.1 | 7 | 964 | |||
GenBank mRNA | AB019397.1 | - | 10 | 1387 | ||
AK302584.1 | - | 10 | 1406 | |||
BC126209.1 | - | 9 | 1330 | 8991925 | ||
BC151237.1 | - | 10 | 1393 | 8860504 | ||
D87448.1 | - | 10 | 1393 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
