siRNA Details
Gene Symbol: CHRM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000739.2 | NP_000730.1 | 4 | 1095 | ||
NM_001006626.1 | NP_001006627.1 | 5 | 1136 | |||
NM_001006627.1 | NP_001006628.1 | 3 | 1269 | |||
NM_001006628.1 | NP_001006629.1 | 3 | 1021 | |||
NM_001006629.1 | NP_001006630.1 | 2 | 952 | |||
NM_001006630.1 | NP_001006631.1 | 4 | 1347 | |||
NM_001006631.1 | NP_001006632.1 | 4 | 1039 | |||
NM_001006632.1 | NP_001006633.1 | 3 | 1017 | |||
XM_011515769.2 | XP_011514071.1 | 3 | 2706 | |||
GenBank mRNA | AF385588.1 | - | 1 | 724 | ||
AF498916.1 | - | 1 | 724 | |||
AK290236.1 | - | 3 | 1248 | |||
AL832585.1 | - | 3 | 1253 | |||
BC095547.1 | - | 1 | 727 | 6519221 | ||
BC106741.1 | - | 1 | 724 | 40017105 | ||
BC106742.1 | - | 1 | 724 | 40017106 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
