siRNA Details
Gene Symbol: Araf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022532.2 | NP_071977.1 | 15 | 1707 | ||
XM_008773036.1 | XP_008771258.1 | 15 | 2449 | |||
GenBank mRNA | X06942.1 | - | 14 | 1674 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
