siRNA Details
Gene Symbol: Prl8a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022537.1 | NP_071982.1 | 2, 3 | 273 | ||
XM_006253909.1 | XP_006253971.1 | 2, 3 | 247 | |||
GenBank mRNA | AB000107.1 | - | 2, 3 | 273 | ||
BC098917.1 | - | 2, 3 | 253 | 7445772 |
