siRNA Details
Gene Symbol: Kcnk10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023096.2 | NP_075584.1 | 7 | 2375 | ||
XM_008764847.1 | XP_008763069.1 | 7 | 3095 | |||
GenBank mRNA | AF385401.1 | - | 7 | 2375 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Kcnk10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023096.2 | NP_075584.1 | 7 | 2375 | ||
XM_008764847.1 | XP_008763069.1 | 7 | 3095 | |||
GenBank mRNA | AF385401.1 | - | 7 | 2375 |