siRNA Details
Gene Symbol: Gucy1a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023956.1 | NP_076446.1 | 2 | 733 | ||
GenBank mRNA | AB096080.1 | - | 2 | 733 | ||
AB097860.1 | - | 2 | 313 | |||
AF109963.2 | - | 2 | 733 |

More Information
Other Species Targeted

Gene Ontology Categories:
