siRNA Details
Gene Symbol: Secisbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024002.1 | NP_076492.1 | 5 | 667 | ||
XM_008771506.1 | XP_008769728.1 | 5 | 714 | |||
GenBank mRNA | AJ251245.1 | - | 5 | 667 | ||
FQ227169.1 | - | 5 | 687 | |||
FQ231169.1 | - | 5 | 689 | |||
FQ232320.1 | - | 5 | 695 | |||
FQ234993.1 | - | 5 | 708 |
