siRNA Details
Gene Symbol: Atf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031018.1 | NP_112280.1 | 5 | 464 | ||
GenBank mRNA | AY724488.1 | - | 336 | |||
M65148.1 | - | 2 | 153 | |||
U38938.1 | - | 5 | 464 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001025093, NM_001284371, NM_001284370, NM_001284372, NM_001284373, and others.
Gene Ontology Categories:
