siRNA Details
Gene Symbol: Ces1e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031565.2 | NP_113753.2 | 2 | 436 | ||
GenBank mRNA | EX493673.1 | - | 2 | 228 | ||
FM063764.1 | - | 2 | 436 | |||
X81395.1 | - | 2 | 301 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
