siRNA Details
Gene Symbol: USH2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007123.5 | NP_009054.5 | 21 | 5310 | ||
GenBank mRNA | AF055580.2 | - | 21 | 5310 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
