siRNA Details
Gene Symbol: Fads1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053445.2 | NP_445897.2 | 2, 3 | 397 | ||
GenBank mRNA | AB052085.1 | - | 2, 3 | 397 | ||
AF320509.1 | - | 2, 3 | 426 | |||
HQ909026.1 | - | 2, 3 | 321 |
