siRNA Details
Gene Symbol: Fads1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053445.2 | NP_445897.2 | 12 | 1529 | ||
XM_006231075.2 | XP_006231137.1 | 12 | 1676 | |||
GenBank mRNA | AB052085.1 | - | 12 | 1529 | ||
AF320509.1 | - | 12 | 1558 |
