siRNA Details
Gene Symbol: Olfm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053573.1 | NP_446025.1 | 3 | 680 | ||
XM_006233875.2 | XP_006233937.1 | 3 | 640 | |||
XM_006233876.2 | XP_006233938.1 | 3 | 708 | |||
XM_006233877.1 | XP_006233939.1 | 3 | 640 | |||
GenBank mRNA | U03414.1 | - | 3 | 543 | ||
U03415.1 | - | 3 | 680 | |||
U03416.1 | - | 3 | 542 | |||
U03417.1 | - | 3 | 680 |

More Information
Other Species Targeted

Gene Ontology Categories:
