siRNA Details
Gene Symbol: Olfm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053573.1 | NP_446025.1 | 4 | 765 | ||
XM_006233875.2 | XP_006233937.1 | 4 | 725 | |||
GenBank mRNA | U03416.1 | - | 4 | 627 | ||
U03417.1 | - | 4 | 765 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001038613, NM_019498, AF028740, D78264, BU512660, and others.
Gene Ontology Categories:
