siRNA Details
Gene Symbol: Dut | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040271.1 | NP_001035361.1 | 1, 2 | 171 | ||
NM_053592.2 | NP_446044.1 | 2, 3 | 291 | |||
XM_008762188.1 | XP_008760410.1 | 2, 3 | 329 | |||
GenBank mRNA | AW530495.1 | - | 1, 2 | 179 | ||
CB325379.1 | - | 1, 2 | 185 | |||
FQ220451.1 | - | 326 | ||||
U64030.2 | - | 2, 3 | 291 |

More Information
Other Species Targeted

Gene Ontology Categories:
