siRNA Details
Gene Symbol: Dffa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053679.2 | NP_446131.2 | 4 | 643 | ||
XM_006239362.2 | XP_006239424.1 | 4 | 664 | |||
GenBank mRNA | AF136601.1 | - | 4 | 628 | ||
BC100067.1 | - | 4 | 643 | 7389030 |
