siRNA Details
Gene Symbol: Kcnk6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053806.2 | NP_446258.2 | 2 | 607 | ||
GenBank mRNA | AF281304.1 | - | 607 | |||
AF281305.1 | - | 3 | 406 |
Gene Symbol: LOC100909725 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003748812.3 | XP_003748860.1 | 1134 |
