siRNA Details
Gene Symbol: Rere | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053885.2 | NP_446337.2 | 16 | 2447 | ||
XM_006239438.2 | XP_006239500.1 | 16 | 2520 | |||
XM_006239441.2 | XP_006239503.1 | 10 | 4427 | |||
XM_008764198.1 | XP_008762420.1 | 16 | 2520 | |||
XM_008764199.1 | XP_008762421.1 | 16 | 2937 | |||
XM_008764200.1 | XP_008762422.1 | 16 | 2520 | |||
XM_008764201.1 | XP_008762423.1 | 15 | 2395 | |||
XM_008764202.1 | XP_008762424.1 | 15 | 1961 | |||
XM_008764203.1 | XP_008762425.1 | 14 | 4088 | |||
GenBank mRNA | U44091.1 | - | 15 | 3256 |
