siRNA Details
Gene Symbol: Pla2g6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005560.1 | NP_001005560.1 | 4, 5 | 710 | ||
NM_001270796.1 | NP_001257725.1 | 5, 6 | 789 | |||
XM_006241997.2 | XP_006242059.1 | 4, 5 | 1532 | |||
XM_006241998.2 | XP_006242060.1 | 4, 5 | 1486 | |||
XM_006241999.2 | XP_006242061.1 | 4, 5 | 1532 | |||
XM_006242000.2 | XP_006242062.1 | 4, 5 | 1211 | |||
XM_006242001.2 | XP_006242063.1 | 5, 6 | 843 | |||
XM_006242002.1 | XP_006242064.1 | 4, 5 | 1079 | |||
XM_006242003.1 | XP_006242065.1 | 5, 6 | 1151 | |||
XM_006242004.2 | XP_006242066.1 | 4, 5 | 772 | |||
XM_006242005.2 | XP_006242067.1 | 4, 5 | 1532 | |||
XM_006242006.2 | XP_006242068.1 | 4, 5 | 1210 | |||
XM_008765756.1 | XP_008763978.1 | 5, 6 | 1119 | |||
XM_008765757.1 | XP_008763979.1 | 4, 5 | 1532 | |||
GenBank mRNA | BC081916.1 | - | 4, 5 | 710 | 7112407 | |
U51898.1 | - | 4, 5 | 1088 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
