siRNA Details
Gene Symbol: Cgrrf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053899.1 | NP_446351.1 | 3 | 436 | ||
GenBank mRNA | BC078763.1 | - | 3 | 452 | 7107668 | |
FQ214200.1 | - | 3 | 347 | |||
FQ218613.1 | - | 3 | 451 | |||
U66471.1 | - | 3 | 436 |
